Assay Details
| Gene Symbol: LOC4341966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787686.2 | XP_015643172.1 | 1 - 2 | 157 | 94 | |
| GenBank mRNA | AK065230.1 | - | 1 - 2 | 137 | 94 | |
| AK099927.1 | - | 1 - 2 | 132 | 94 | ||
| AK105454.1 | - | 1 - 2 | 138 | 94 | ||
| AK119234.1 | - | 1 - 2 | 136 | 94 | ||
| AK242947.1 | - | 1278 | 94 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787686.2 transcripts.
Set Membership: |
|

LOC4341966
Genomic Map