Assay Details
| Gene Symbol: LOC4341966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787686.2 | XP_015643172.1 | 2 - 3 | 229 | 81 | |
| GenBank mRNA | AK065230.1 | - | 2 - 3 | 209 | 81 | |
| AK099927.1 | - | 2 - 3 | 204 | 81 | ||
| AK105454.1 | - | 2 - 3 | 210 | 81 | ||
| AK119234.1 | - | 2 - 3 | 208 | 81 | ||
| AK242947.1 | - | 1350 | 81 | |||
| D12627.1 | - | 1 - 2 | 82 | 81 | ||
More Information
Set Membership: |
|

LOC4341966
Genomic Map