Assay Details
| Gene Symbol: LOC4342193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789253.2 | XP_015644739.1 | 8 - 9 | 1344 | 82 | |
| GenBank mRNA | AK063949.1 | - | 2 - 3 | 215 | 82 | |
More Information
Set Membership: |
|

LOC4342193
Genomic Map