Assay Details
| Gene Symbol: LOC4342193 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789253.2 | XP_015644739.1 | 9 - 10 | 1470 | 78 | |
| GenBank mRNA | AK063949.1 | - | 3 - 4 | 341 | 78 | |
More Information
Set Membership: |
|

LOC4342193
Genomic Map