Assay Details
Gene Symbol: LOC4342207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791220.2 | XP_015646706.1 | 7 - 8 | 1075 | 131 | |
GenBank mRNA | AK241298.1 | - | 7 - 8 | 1038 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() |
