Assay Details
| Gene Symbol: LOC4342469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791807.2 | XP_015647293.1 | 25 - 26 | 2929 | 79 | |
| XM_015791808.2 | XP_015647294.1 | 25 - 26 | 2864 | 79 | ||
| GenBank mRNA | AK241926.1 | - | 4 - 5 | 367 | 79 | |
More Information
Set Membership: |
|

LOC4342469
Genomic Map