Assay Details
| Gene Symbol: LOC4342469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791807.2 | XP_015647293.1 | 28 - 29 | 3147 | 114 | |
| XM_015791808.2 | XP_015647294.1 | 28 - 29 | 3082 | 114 | ||
| GenBank mRNA | AK241926.1 | - | 7 - 8 | 585 | 113 | |
More Information
Set Membership: |
|

LOC4342469
Genomic Map