Assay Details
| Gene Symbol: LOC4342635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791930.1 | XP_015647416.1 | 10 - 11 | 1277 | 98 | |
| XM_015791931.1 | XP_015647417.1 | 8 - 9 | 992 | 98 | ||
| GenBank mRNA | AK070547.1 | - | 9 - 10 | 815 | 98 | |
More Information
Set Membership: |
|

LOC4342635
Genomic Map