Assay Details
| Gene Symbol: LOC4342694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791393.2 | XP_015646879.1 | 1 - 2 | 2636 | 60 | |
| GenBank mRNA | AK111823.1 | - | 1 - 2 | 2597 | 60 | |
More Information
Set Membership: |
|

LOC4342694
Genomic Map