Assay Details
| Gene Symbol: LOC4342819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789740.2 | XP_015645226.1 | 15 - 16 | 1609 | 126 | |
| GenBank mRNA | AK121036.1 | - | 15 - 16 | 1616 | 126 | |
More Information
Set Membership: |
|

LOC4342819
Genomic Map