Assay Details
| Gene Symbol: LOC9271299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791212.2 | XP_015646698.1 | 19 - 20 | 2201 | 92 | |
| XM_015791213.2 | XP_015646699.1 | 19 - 20 | 2198 | 92 | ||
| GenBank mRNA | AK102722.1 | - | 17 - 18 | 2317 | 92 | |
More Information
Set Membership: |
|

LOC9271299
Genomic Map