Assay Details
| Gene Symbol: LOC9271299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791212.2 | XP_015646698.1 | 22 - 23 | 2475 | 89 | |
| XM_015791213.2 | XP_015646699.1 | 22 - 23 | 2472 | 89 | ||
| GenBank mRNA | AK102722.1 | - | 20 - 21 | 2591 | 89 | |
More Information
Set Membership: |
|

LOC9271299
Genomic Map