Assay Details
| Gene Symbol: LOC9271299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791212.2 | XP_015646698.1 | 5 - 6 | 599 | 81 | |
| XM_015791213.2 | XP_015646699.1 | 5 - 6 | 599 | 81 | ||
| GenBank mRNA | AK058401.1 | - | 196 | 81 | ||
| AK102722.1 | - | 5 - 6 | 514 | 81 | ||
More Information
Set Membership: |
|

LOC9271299
Genomic Map