Assay Details
| Gene Symbol: LOC9271299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791213.2 | XP_015646699.1 | 6 - 7 | 679 | 77 | |
| GenBank mRNA | AK058401.1 | - | 276 | 77 | ||
| AK102722.1 | - | 6 - 7 | 594 | 77 | ||
More Information
Set Membership: |
|

LOC9271299
Genomic Map