Assay Details
Gene Symbol: LOC4343610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792290.2 | XP_015647776.1 | 1 - 2 | 255 | 80 | |
GenBank mRNA | AK067575.1 | - | 1 - 2 | 232 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792290.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
