Assay Details
| Gene Symbol: LOC4343653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789406.2 | XP_015644892.1 | 2 | 1210 | 86 | |
| GenBank mRNA | AK062388.1 | - | 1 | 450 | 86 | |
| HQ324818.1 | - | 2 | 1148 | 86 | ||
More Information
Set Membership: |
|

LOC4343653
Genomic Map