Assay Details
Gene Symbol: LOC4343765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791722.2 | XP_015647208.1 | 1 | 715 | 62 | |
GenBank mRNA | AY395294.1 | - | 1 | 553 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
