Assay Details
Gene Symbol: LOC4343787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790682.2 | XP_015646168.1 | 2 - 3 | 401 | 116 | |
GenBank mRNA | AK121652.1 | - | 1 - 2 | 286 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
