Assay Details
| Gene Symbol: LOC4343787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790682.2 | XP_015646168.1 | 3 - 4 | 512 | 78 | |
| GenBank mRNA | AK121652.1 | - | 2 - 3 | 397 | 78 | |
More Information
Set Membership: |
|

LOC4343787
Genomic Map