Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 11 - 12 | 1190 | 82 | |
| GenBank mRNA | AK066811.1 | - | 3 - 4 | 292 | 82 | |
| AK069020.1 | - | 11 - 12 | 1152 | 82 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map