Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 12 - 13 | 1272 | 73 | |
| GenBank mRNA | AK066811.1 | - | 4 - 5 | 374 | 73 | |
| AK069020.1 | - | 12 - 13 | 1234 | 73 | ||
| AK104850.1 | - | 1 - 2 | 67 | 73 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map