Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 14 - 15 | 1400 | 106 | |
| GenBank mRNA | AK066811.1 | - | 6 - 7 | 502 | 106 | |
| AK069020.1 | - | 14 - 15 | 1362 | 106 | ||
| AK104850.1 | - | 3 - 4 | 105 | |||
More Information
Set Membership: |
|

LOC4344031
Genomic Map