Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 15 - 16 | 1530 | 144 | |
| GenBank mRNA | AK066811.1 | - | 7 - 8 | 632 | 144 | |
| AK069020.1 | - | 15 - 16 | 1492 | 144 | ||
| AK104850.1 | - | 4 - 5 | 324 | 144 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map