Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 16 - 17 | 1595 | 110 | |
| GenBank mRNA | AK066811.1 | - | 8 - 9 | 697 | 110 | |
| AK069020.1 | - | 16 - 17 | 1557 | 110 | ||
| AK104850.1 | - | 5 - 6 | 389 | 110 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map