Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 19 - 20 | 1813 | 130 | |
| GenBank mRNA | AK066811.1 | - | 11 - 12 | 915 | 130 | |
| AK069020.1 | - | 19 - 20 | 1775 | 130 | ||
| AK104850.1 | - | 8 - 9 | 607 | 130 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map