Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 21 - 22 | 2045 | 72 | |
| GenBank mRNA | AK066811.1 | - | 13 - 14 | 1147 | 72 | |
| AK069020.1 | - | 21 - 22 | 2007 | 72 | ||
| AK104850.1 | - | 10 - 11 | 839 | 72 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map