Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 23 - 24 | 2148 | 73 | |
| GenBank mRNA | AK066811.1 | - | 15 - 16 | 1250 | 73 | |
| AK069020.1 | - | 23 - 24 | 2110 | 73 | ||
| AK104850.1 | - | 12 - 13 | 942 | 73 | ||
More Information
Set Membership: |
|

LOC4344031
Genomic Map