Assay Details
| Gene Symbol: LOC4344031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790638.2 | XP_015646124.1 | 5 - 6 | 702 | 92 | |
| GenBank mRNA | AK069020.1 | - | 5 - 6 | 664 | 92 | |
More Information
Set Membership: |
|

LOC4344031
Genomic Map