Assay Details
| Gene Symbol: LOC4344043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789090.2 | XP_015644576.1 | 28 - 29 | 5537 | 109 | |
| XM_015789091.2 | XP_015644577.1 | 28 - 29 | 5528 | 109 | ||
| GenBank mRNA | AK060298.1 | - | 6 - 7 | 718 | 109 | |
More Information
Set Membership: |
|

LOC4344043
Genomic Map