Assay Details
| Gene Symbol: LOC4344043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789090.2 | XP_015644576.1 | 29 - 30 | 5612 | 125 | |
| XM_015789091.2 | XP_015644577.1 | 29 - 30 | 5603 | 125 | ||
| GenBank mRNA | AK060298.1 | - | 7 - 8 | 793 | 125 | |
More Information
Set Membership: |
|

LOC4344043
Genomic Map