Assay Details
| Gene Symbol: LOC4344171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789297.1 | XP_015644783.1 | 2 - 3 | 337 | 66 | |
| GenBank mRNA | AK061060.1 | - | 2 - 3 | 335 | 66 | |
More Information
Set Membership: |
|

LOC4344171
Genomic Map