Assay Details
| Gene Symbol: LOC4344300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791282.2 | XP_015646768.1 | 4 - 5 | 426 | 85 | |
| GenBank mRNA | AK066982.1 | - | 4 - 5 | 388 | 85 | |
More Information
Set Membership: |
|

LOC4344300
Genomic Map