Assay Details
| Gene Symbol: LOC4344345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791264.2 | XP_015646750.1 | 3 - 4 | 841 | 94 | |
| GenBank mRNA | AK073043.1 | - | 3 - 4 | 620 | 94 | |
More Information
Set Membership: |
|

LOC4344345
Genomic Map