Assay Details
| Gene Symbol: LOC4344373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791939.2 | XP_015647425.1 | 2 - 3 | 833 | 76 | |
| GenBank mRNA | AK058311.1 | - | 2 - 3 | 829 | 76 | |
| AK070759.1 | - | 1 - 2 | 130 | 76 | ||
| AK099264.1 | - | 2 - 3 | 830 | 76 | ||
| AK105861.1 | - | 2 - 3 | 829 | 76 | ||
| D17789.1 | - | 2 - 3 | 797 | 76 | ||
More Information
Set Membership: |
|

LOC4344373
Genomic Map