Assay Details
| Gene Symbol: LOC4344382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790690.1 | XP_015646176.1 | 1 | 651 | 79 | |
| GenBank mRNA | AB111050.1 | - | 1 | 538 | 79 | |
More Information
Set Membership: |
|

LOC4344382
Genomic Map