Assay Details
| Gene Symbol: LOC4344541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794725.1 | XP_015650211.1 | 1 - 2 | 737 | 91 | |
| XM_015794727.1 | XP_015650213.1 | 1 - 2 | 737 | 91 | ||
| GenBank mRNA | AB017428.1 | - | 1 - 2 | 728 | 91 | |
| AK059815.1 | - | 1 - 2 | 742 | 91 | ||
| AK099077.1 | - | 1 - 2 | 739 | 91 | ||
| AK104303.1 | - | 1 - 2 | 739 | 91 | ||
| AK104304.1 | - | 1 - 2 | 740 | 91 | ||
| AK104421.1 | - | 1 - 2 | 740 | 91 | ||
More Information
Set Membership: |
|

LOC4344541
Genomic Map