Assay Details
| Gene Symbol: LOC4344564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794644.2 | XP_015650130.1 | 1 - 2 | 200 | 67 | |
| GenBank mRNA | AK071097.1 | - | 1 - 2 | 88 | 67 | |
| AK103777.1 | - | 1 - 2 | 55 | 67 | ||
| EF122472.1 | - | 1 - 2 | 73 | 67 | ||
| GQ848031.1 | - | 1 - 2 | 71 | 67 | ||
| JF969200.1 | - | 1 - 2 | 51 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794644.2 transcripts.
Set Membership: |
|

LOC4344564
Genomic Map