Assay Details
| Gene Symbol: LOC4344564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794644.2 | XP_015650130.1 | 4 - 5 | 432 | 71 | |
| GenBank mRNA | AK071097.1 | - | 4 - 5 | 320 | 71 | |
| AK103777.1 | - | 4 - 5 | 287 | 71 | ||
| EF122472.1 | - | 4 - 5 | 305 | 71 | ||
| GQ848031.1 | - | 4 - 5 | 303 | 71 | ||
| JF969200.1 | - | 4 - 5 | 283 | 71 | ||
| JN848809.1 | - | 3 - 4 | 243 | 71 | ||
More Information
Set Membership: |
|

LOC4344564
Genomic Map