Assay Details
| Gene Symbol: LOC4344564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794644.2 | XP_015650130.1 | 7 - 8 | 745 | 81 | |
| GenBank mRNA | AK071097.1 | - | 7 - 8 | 633 | 81 | |
| AK103777.1 | - | 7 - 8 | 600 | 81 | ||
| EF122472.1 | - | 7 - 8 | 618 | 81 | ||
| GQ848031.1 | - | 7 - 8 | 616 | 81 | ||
| JF969200.1 | - | 7 - 8 | 596 | 81 | ||
| JN848809.1 | - | 6 - 7 | 556 | 81 | ||
More Information
Set Membership: |
|

LOC4344564
Genomic Map