Assay Details
| Gene Symbol: LOC4344564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794644.2 | XP_015650130.1 | 9 - 10 | 975 | 66 | |
| GenBank mRNA | AK071097.1 | - | 9 - 10 | 863 | 66 | |
| AK103777.1 | - | 9 - 10 | 830 | 66 | ||
| EF122472.1 | - | 9 - 10 | 848 | 66 | ||
| GQ848031.1 | - | 9 - 10 | 846 | 66 | ||
| JF969200.1 | - | 9 - 10 | 826 | 66 | ||
| JN848809.1 | - | 8 - 9 | 786 | 66 | ||
More Information
Set Membership: |
|

LOC4344564
Genomic Map