Assay Details
| Gene Symbol: LOC9266786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792644.2 | XP_015648130.1 | 5 - 6 | 675 | 143 | |
| XM_015792645.2 | XP_015648131.1 | 5 - 6 | 432 | 143 | ||
| GenBank mRNA | AK242866.1 | - | 1921 | 143 | ||
| Gene Symbol: LOC4323897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288851.1 | - | 831 | 143 | ||
More Information
Set Membership: |
|

LOC9266786
Genomic Map