Assay Details
| Gene Symbol: LOC4345209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795406.2 | XP_015650892.1 | 7 - 8 | 845 | 151 | |
| GenBank mRNA | AK061379.1 | - | 6 - 7 | 654 | 151 | |
| AK099389.1 | - | 6 - 7 | 620 | 151 | ||
| AK101640.1 | - | 6 - 7 | 620 | 151 | ||
More Information
Set Membership: |
|

LOC4345209
Genomic Map