Assay Details
| Gene Symbol: LOC4345254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795515.2 | XP_015651001.1 | 1 - 2 | 294 | 62 | |
| GenBank mRNA | AK121802.1 | - | 1 - 2 | 107 | 62 | |
More Information
Set Membership: |
|

LOC4345254
Genomic Map