Assay Details
| Gene Symbol: LOC4345340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795475.2 | XP_015650961.1 | 2 - 3 | 368 | 84 | |
| GenBank mRNA | AK059644.1 | - | 2 - 3 | 326 | 84 | |
| AK103873.1 | - | 2 - 3 | 369 | 84 | ||
More Information
Set Membership: |
|

LOC4345340
Genomic Map