Assay Details
| Gene Symbol: LOC4345411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792695.2 | XP_015648181.1 | 1 - 2 | 188 | 125 | |
| GenBank mRNA | AK070541.1 | - | 1 - 2 | 149 | 125 | |
| AK104158.1 | - | 1 - 2 | 107 | 125 | ||
| AK104511.1 | - | 1 - 2 | 122 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792695.2 transcripts.
Set Membership: |
|

LOC4345411
Genomic Map