Assay Details
| Gene Symbol: LOC4345411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792695.2 | XP_015648181.1 | 2 - 3 | 299 | 152 | |
| GenBank mRNA | AK070541.1 | - | 2 - 3 | 260 | 152 | |
| AK104511.1 | - | 2 - 3 | 233 | 152 | ||
More Information
Set Membership: |
|

LOC4345411
Genomic Map