Assay Details
| Gene Symbol: LOC4345474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794668.2 | XP_015650154.1 | 4 - 5 | 2189 | 154 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4345474| Gene Symbol: LOC4345474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794668.2 | XP_015650154.1 | 4 - 5 | 2189 | 154 | |
Set Membership: |
|