Assay Details
Gene Symbol: LOC4346491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756868.2 | XP_015612354.1 | 6 - 7 | 1603 | 81 | |
XM_015756870.2 | XP_015612356.1 | 6 - 7 | 1603 | 81 | ||
GenBank mRNA | AB096014.1 | - | 6 - 7 | 1485 | 81 | |
AB120431.1 | - | 6 - 7 | 1575 | 81 | ||
AK059162.1 | - | 1 - 2 | 80 | 81 | ||
AK066809.1 | - | 6 - 7 | 1576 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
