Assay Details
| Gene Symbol: LOC4346585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756599.2 | XP_015612085.1 | 3 - 4 | 3086 | 58 | |
| XM_015756602.1 | XP_015612088.1 | 1 - 2 | 280 | 58 | ||
| GenBank mRNA | AK111610.1 | - | 2 - 3 | 3153 | 58 | |
More Information
Set Membership: |
|

LOC4346585
Genomic Map