Assay Details
| Gene Symbol: LOC4346843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756955.1 | XP_015612441.1 | 6 - 7 | 780 | 146 | |
| GenBank mRNA | AK099826.1 | - | 6 - 7 | 786 | 146 | |
More Information
Set Membership: |
|

LOC4346843
Genomic Map